missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone 99.0+%, TCI America™

Supplier:  TCI America H0266500G

 View more versions of this product

Catalog No. H0266500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Hydroxy-4-methoxybenzophenone
63°C
MFCD00008387
3077
DXGLGDHPHMLXJC-UHFFFAOYSA-N
(2-hydroxy-4-methoxyphenyl)-phenylmethanone
4632
228.25
Crystalline Powder
131-57-7
C14H12O3
500 g
oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O
228.247
CHEBI:34283
≥99.0% (GC)

Chemical Identifiers

131-57-7
228.247
DXGLGDHPHMLXJC-UHFFFAOYSA-N
4632
(2-hydroxy-4-methoxyphenyl)-phenylmethanone
C14H12O3
MFCD00008387
oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
CHEBI:34283
COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0130

RTECSNumber : DJ1575000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions