missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone 99.0+%, TCI America™

Catalog No. p-7130451
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H026625G 25 g
H0266500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H026625G Supplier TCI America Supplier No. H026625G
Only null left

Chemical Identifiers

CAS 131-57-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00008387
InChI Key DXGLGDHPHMLXJC-UHFFFAOYSA-N
Synonym oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
PubChem CID 4632
ChEBI CHEBI:34283
IUPAC Name (2-hydroxy-4-methoxyphenyl)-phenylmethanone
SMILES COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

Specifications

Melting Point 63°C
Quantity 25 g
UN Number 3077
Formula Weight 228.25
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone
EINECSNumber (4)-0130
RTECSNumber DJ1575000
TSCA Yes