Learn More
2-Hydroxy-4-methoxybenzophenone, 98+%
CAS: 131-57-7 | C14H12O3 | 228.247 g/mol
Supplier: Thermo Scientific Chemicals A1766218
Description
2-Hydroxy-4-methoxybenzophenone is used as a broad-band filter in sunscreens cosmetic products in order to protect the skin from the deleterious effects of the sun. It acts as a reagent in the gravimetric determination of copper(II). Further, it is used in the preparation of poly[(2-hydroxy-4-methoxybenzophenone) propylene], which is a polymeric ligand useful for the synthesis of metal/ligand polychelates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxy-4-methoxybenzophenone is used as a broad-band filter in sunscreens cosmetic products in order to protect the skin from the deleterious effects of the sun. It acts as a reagent in the gravimetric determination of copper(II). Further, it is used in the preparation of poly[(2-hydroxy-4-methoxybenzophenone) propylene], which is a polymeric ligand useful for the synthesis of metal/ligand polychelates.
Solubility
Insoluble in water.
Notes
Store in a cool place. Incompatible with bases, acid chlorides, acid anhydrides and oxidizing agents.
Specifications
| 2-Hydroxy-4-methoxybenzophenone | |
| 62°C to 65°C | |
| ∼150°C (5mmHg) | |
| C14H12O3 | |
| 50 g | |
| 14,6954 | |
| Insoluble in water. | |
| COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O | |
| 228.247 | |
| CHEBI:34283 | |
| ≥98% |
| 131-57-7 | |
| 1.3 | |
| 216°C (420°F) | |
| MFCD00008387 | |
| 1913145 | |
| oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex | |
| DXGLGDHPHMLXJC-UHFFFAOYSA-N | |
| (2-hydroxy-4-methoxyphenyl)-phenylmethanone | |
| 4632 | |
| 228.25 |
Chemical Identifiers
| 131-57-7 | |
| 228.247 | |
| DXGLGDHPHMLXJC-UHFFFAOYSA-N | |
| 4632 | |
| (2-hydroxy-4-methoxyphenyl)-phenylmethanone |
| C14H12O3 | |
| MFCD00008387 | |
| oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex | |
| CHEBI:34283 | |
| COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 205-031-5
RTECSNumber : DJ1575000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only