missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxybenzophenone, 98+%

Catalog No. p-7023776
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1766218 50 g
AAA1766230 250 g
AAA176620B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1766218 Supplier Thermo Scientific Chemicals Supplier No. A1766218

CAS: 131-57-7 | C14H12O3 | 228.247 g/mol

2-Hydroxy-4-methoxybenzophenone is used as a broad-band filter in sunscreens cosmetic products in order to protect the skin from the deleterious effects of the sun. It acts as a reagent in the gravimetric determination of copper(II). Further, it is used in the preparation of poly[(2-hydroxy-4-methoxybenzophenone) propylene], which is a polymeric ligand useful for the synthesis of metal/ligand polychelates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-4-methoxybenzophenone is used as a broad-band filter in sunscreens cosmetic products in order to protect the skin from the deleterious effects of the sun. It acts as a reagent in the gravimetric determination of copper(II). Further, it is used in the preparation of poly[(2-hydroxy-4-methoxybenzophenone) propylene], which is a polymeric ligand useful for the synthesis of metal/ligand polychelates.

Solubility
Insoluble in water.

Notes
Store in a cool place. Incompatible with bases, acid chlorides, acid anhydrides and oxidizing agents.

Chemical Identifiers

CAS 131-57-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00008387
InChI Key DXGLGDHPHMLXJC-UHFFFAOYSA-N
Synonym oxybenzone, 2-hydroxy-4-methoxybenzophenone, benzophenone-3, oxybenzon, 4-methoxy-2-hydroxybenzophenone, 2-benzoyl-5-methoxyphenol, oxibenzona, oxybenzonum, 2-hydroxy-4-methoxyphenyl phenyl methanone, anuvex
PubChem CID 4632
ChEBI CHEBI:34283
IUPAC Name (2-hydroxy-4-methoxyphenyl)-phenylmethanone
SMILES COC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)O

Specifications

Melting Point 62°C to 65°C
Density 1.3
Boiling Point ∼150°C (5mmHg)
Flash Point 216°C (420°F)
Quantity 50 g
Beilstein 1913145
Merck Index 14,6954
Solubility Information Insoluble in water.
Formula Weight 228.25
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxy-4-methoxybenzophenone
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 205-031-5
RTECSNumber DJ1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only