missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-4,4',6'-trimethoxychalcone 98.0+%, TCI America™

Catalog No. H15715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H15715G 5 g
H15711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H15715G Supplier TCI America Supplier No. H15715G
Only null left

Chemical Identifiers

CAS 3420-72-2
Molecular Formula C18H18O5
Molecular Weight (g/mol) 314.337
MDL Number MFCD00017174
InChI Key CGIBCVBDFUTMPT-RMKNXTFCSA-N
Synonym Flavokawain A
PubChem CID 5355469
IUPAC Name (E)-1-(2-hydroxy-4,6-dimethoxyphenyl)-3-(4-methoxyphenyl)prop-2-en-1-one
SMILES COC1=CC=C(C=C1)C=CC(=O)C2=C(C=C(C=C2OC)OC)O

Specifications

Melting Point 114°C
Quantity 5 g
Formula Weight 314.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Hydroxy-4,4′,6′-trimethoxychalcone