missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxy-3-phenylpropiophenone 98.0+%, TCI America™

Numéro de catalogue. H131325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H131325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. H131325G Fournisseur TCI America Code fournisseur H131325G
Il en reste null

Identifiants chimiques

CAS 3516-95-8
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00002221
InChI Key JCPGMXJLFWGRMZ-UHFFFAOYSA-N
Synonym 2-Hydroxyphenyl Phenethyl Ketone, 2-Hydroxyphenyl 2-Phenylethyl Ketone
PubChem CID 77052
IUPAC Name 1-(2-hydroxyphenyl)-3-phenylpropan-1-one
SMILES C1=CC=C(C=C1)CCC(=O)C2=CC=CC=C2O

Spécifications

Melting Point 30°C
Color White-Yellow
Boiling Point 158°C
Quantity 25 g
Formula Weight 226.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2′-Hydroxy-3-phenylpropiophenone