missing translation for 'onlineSavingsMsg'
Learn More

2-Hexyn-1-ol, 97%

Catalog No. AAA1333914
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1333914 25 g
AAA1333906 5 g
AAA1333909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1333914 Supplier Thermo Scientific Chemicals Supplier No. A1333914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 764-60-3 | C6H10O | 98.15 g/mol

2-Hexyn-1-ol, is an important organic primary and secondary intermediate. It can be used in Pharmaceuticals, Agrochemicals and Dyestuff industry. It also acts as a chemical solvent in fine chemicals

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hexyn-1-ol, is an important organic primary and secondary intermediate. It can be used in Pharmaceuticals, Agrochemicals and Dyestuff industry. It also acts as a chemical solvent in fine chemicals

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Avoid temperature and direct sunlight.

Chemical Identifiers

CAS 764-60-3
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.15
MDL Number MFCD00039540
InChI Key BTDWSZJDLLLTMI-UHFFFAOYSA-N
Synonym 2-hexyn-1-ol, pubchem13064, acmc-209p3i, ksc380a2d, 2-hexyn-1-ol,
PubChem CID 69818
IUPAC Name hex-2-yn-1-ol
SMILES CCCC#CCO

Specifications

Density 0.897
Boiling Point 167°C
Flash Point 68°C (154°F)
Refractive Index 1.454
Linear Formula CH3(CH2)2C≡CCH2OH
Quantity 25 g
Beilstein 1736274
Solubility Information It is soluble in water.
Formula Weight 98.15
Percent Purity 97%
Chemical Name or Material 2-Hexyn-1-ol
Show More Show Less
Hazard Category H227-H302
Hazard Statement GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 212-128-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only