missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hexyldecanoic Acid 98.0+%, TCI America™
$56.95 - $317.07
Chemical Identifiers
| CAS | 25354-97-6 |
|---|---|
| Molecular Formula | C16H32O2 |
| Molecular Weight (g/mol) | 256.43 |
| MDL Number | MFCD00059651 |
| InChI Key | JMOLZNNXZPAGBH-UHFFFAOYSA-N |
| Synonym | decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16 |
| PubChem CID | 32912 |
| IUPAC Name | 2-hexyldecanoic acid |
| SMILES | CCCCCCCCC(CCCCCC)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
H050725ML
|
N/A | 25 mL |
Each for $56.95
|
|
|||||
|
H0507100ML
|
N/A | 100 mL |
Each for $121.51
|
|
|||||
|
H0507500ML
|
N/A | 500 mL |
Each for $317.07
|
|
|||||
Chemical Identifiers
| 25354-97-6 | |
| 256.43 | |
| JMOLZNNXZPAGBH-UHFFFAOYSA-N | |
| 32912 | |
| CCCCCCCCC(CCCCCC)C(=O)O |
| C16H32O2 | |
| MFCD00059651 | |
| decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16 | |
| 2-hexyldecanoic acid |
Specifications
| 25354-97-6 | |
| 150°C | |
| MFCD00059651 | |
| JMOLZNNXZPAGBH-UHFFFAOYSA-N | |
| 2-hexyldecanoic acid | |
| 32912 | |
| ≥98.0% (GC) | |
| 2-Hexyldecanoic Acid |
| Yellow | |
| C16H32O2 | |
| decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16 | |
| CCCCCCCCC(CCCCCC)C(=O)O | |
| 256.43 | |
| 256.43 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : HD9750000
TSCA : Yes