missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hexyldecanoic Acid 98.0+%, TCI America™
Supplier: TCI America H0507100ML
| Quantity | 100 mL |
|---|
Chemical Identifiers
| 25354-97-6 | |
| 256.43 | |
| JMOLZNNXZPAGBH-UHFFFAOYSA-N | |
| 32912 | |
| CCCCCCCCC(CCCCCC)C(=O)O |
| C16H32O2 | |
| MFCD00059651 | |
| decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16 | |
| 2-hexyldecanoic acid |
Specifications
| 2-Hexyldecanoic Acid | |
| Yellow | |
| C16H32O2 | |
| MFCD00059651 | |
| JMOLZNNXZPAGBH-UHFFFAOYSA-N | |
| 2-hexyldecanoic acid | |
| 32912 | |
| ≥98.0% (GC) |
| 25354-97-6 | |
| 150°C | |
| 100 mL | |
| decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16 | |
| CCCCCCCCC(CCCCCC)C(=O)O | |
| 256.43 | |
| 256.43 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0608
RTECSNumber : HD9750000
TSCA : Yes