accessibility menu, dialog, popup

UserName

H3CCH3OHO

CAS RN 25354-97-6

IUPAC Name: 2-hexyldecanoic acid
Synonyms: hexyldecanoic acid
Molecular Weight (g/mol): 256.43
Molecular Formula: C16H32O2
InChi Key: JMOLZNNXZPAGBH-UHFFFAOYNA-N
SMILES: CCCCCCCCC(CCCCCC)C(O)=O
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)