missing translation for 'onlineSavingsMsg'
Learn More

2-Hexyldecanoic Acid 98.0+%, TCI America™

Catalog No. H050725ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H050725ML 25 mL
H0507100ML 100 mL
H0507500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H050725ML Supplier TCI America Supplier No. H050725ML

Chemical Identifiers

CAS 25354-97-6
Molecular Formula C16H32O2
Molecular Weight (g/mol) 256.43
MDL Number MFCD00059651
InChI Key JMOLZNNXZPAGBH-UHFFFAOYSA-N
Synonym decanoic acid, 2-hexyl, hexyldecanoic acid, c16 guerbet fatty acid, 2-hexyldecansaeure, 7-pentadecanecarboxylic acid, 2-hexyldecansaeure german, 2-hexyl-decanoic acid, 2-n-hexyldecanoic acid, 2-hexyldecanoicacid, isocarb 16
PubChem CID 32912
IUPAC Name 2-hexyldecanoic acid
SMILES CCCCCCCCC(CCCCCC)C(=O)O

Specifications

Color Yellow
Boiling Point 150°C
Quantity 25 mL
Formula Weight 256.43
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Hexyldecanoic Acid
EINECSNumber (2)-0608
RTECSNumber HD9750000
TSCA Yes