missing translation for 'onlineSavingsMsg'
Learn More

2-Hexyl-4-pentynoic Acid 95.0+%, TCI America™

Numéro de catalogue. H09645G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
H09645G 5 g
H096425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. H09645G Fournisseur TCI America Code fournisseur H09645G
Il en reste null

Identifiants chimiques

CAS 96017-59-3
Molecular Formula C11H18O2
Molecular Weight (g/mol) 182.26
MDL Number MFCD00946863
InChI Key DUQSBRQHALCSLC-UHFFFAOYNA-N
Synonym 2-Propargyloctanoic Acid, 2-(2-Propynyl)octanoic Acid
PubChem CID 175664
IUPAC Name 2-(prop-2-yn-1-yl)octanoic acid
SMILES CCCCCCC(CC#C)C(O)=O

Spécifications

Color Yellow
Quantity 5 g
Formula Weight 182.26
Percent Purity ≥95.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Hexyl-4-pentynoic Acid