missing translation for 'onlineSavingsMsg'
Learn More

2-Hexanone, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018963
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018963 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018963 Supplier Merck Emd Millipore Supplier No. 024735ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2-Hexanone (MBK, 2-Oxohexane) is a volatile organic compound, that exists as a clear, colorless liquid with a sharp odor. It dissolves very easily in water, and can evaporate easily into the air as a vapor. It is occasionally found as a volatile component of normal human biofluids. It was used as an industrial hexacarbon solvent, but is no longer used or made owing to its neurotoxic nature leading to various human diseases. Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care

Specifications

CAS 591-78-6
Vapor Pressure 10 mmHg (39°C)
Density 0.812 g/mL (at 25°C)
Flash Point 23°C
Linear Formula CH3(CH2)3 COCH3
MDL Number MFCD00009482
Melting Point -57°C
Boiling Point 127°C
Synonym Butyl methyl ketone
Type Analytical Standard
Quantity 5 mL
Formula Weight 100.16
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less