missing translation for 'onlineSavingsMsg'
Learn More

2-Heptyn-1-ol 98.0+%, TCI America™

Catalog No. H04565ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04565ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04565ML Supplier TCI America Supplier No. H04565ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1002-36-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00039541
InChI Key OGDYROFIIXDTQK-UHFFFAOYSA-N
Synonym 2-heptyn-1-ol, 2-heptyne-1-ol, 2-heptynol, pubchem13065, acmc-2097no, hoch2c=c ch2 3ch3
PubChem CID 535111
IUPAC Name hept-2-yn-1-ol
SMILES CCCCC#CCO

Specifications

Color Yellow
Boiling Point 114°C
Quantity 5 mL
Formula Weight 112.17
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Heptyn-1-ol