missing translation for 'onlineSavingsMsg'
Learn More

2-Furylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. F03941G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F03941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F03941G Supplier TCI America Supplier No. F03941G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2-Furylboronic Acid (contains varying amounts of Anhydride)
CAS 13331-23-2
Molecular Formula C4H5BO3
Quantity 1 g
MDL Number MFCD00799544
Synonym 2-furanboronic acid, furan-2-boronic acid, 2-furylboronic acid, 2-furanylboronic acid, furan-2-ylboranediol, furan 2-boronic acid, 2-furan boronic acid, furan-2-yl boronic acid, boronic acid, furanyl, 2furylboronic acid
InChI Key PZJSZBJLOWMDRG-UHFFFAOYSA-N
SMILES B(C1=CC=CO1)(O)O
IUPAC Name furan-2-ylboronic acid
Molecular Weight (g/mol) 111.891
PubChem CID 2734357
Formula Weight 111.89
Physical Form Crystalline Powder
Show More Show Less