missing translation for 'onlineSavingsMsg'
Learn More

2-Furoyltrifluoroacetone, TCI America™

Catalog No. F00835G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F00835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F00835G Supplier TCI America Supplier No. F00835G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 326-90-9
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD00020935
InChI Key OWLPCALGCHDBCN-UHFFFAOYSA-N
Synonym 4,4,4-trifluoro-1-furan-2-yl butane-1,3-dione, 2-furoyltrifluoroacetone, furoyltrifluoroacetone, 4,4,4-trifluoro-1-2-furyl-1,3-butanedione, perfluoroacetyl 2-furoyl methane, furonyltrifluoroacetone, 1,3-butanedione, 4,4,4-trifluoro-1-2-furanyl, 4,4,4-trifluoro-1-2-furyl butane-1,3-dione, 2-furoyl trifluoro acetone, 4,4,4-trifluoro-1-2-furanyl-1,3-butanedione
PubChem CID 67594
IUPAC Name 4,4,4-trifluoro-1-(furan-2-yl)butane-1,3-dione
SMILES FC(F)(F)C(=O)CC(=O)C1=CC=CO1

Specifications

Boiling Point 84°C
Quantity 5 g
Formula Weight 206.12
Physical Form Crystalline Lumps
Chemical Name or Material 2-Furoyltrifluoroacetone