missing translation for 'onlineSavingsMsg'
Learn More

2-Furoyl chloride, 98+%

Catalog No. AC165381000
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC165381000 100 mL Glass bottle
AC165380250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC165381000 Supplier Thermo Scientific Chemicals Supplier No. 165381000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 527-69-5 | C5H3ClO2 | 130.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 527-69-5
Molecular Formula C5H3ClO2
Molecular Weight (g/mol) 130.53
MDL Number MFCD00003228
InChI Key OFTKFKYVSBNYEC-UHFFFAOYSA-N
Synonym 2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride
PubChem CID 68242
IUPAC Name furan-2-carbonyl chloride
SMILES C1=COC(=C1)C(=O)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -2°C
Color Yellow to Brown
Density 1.3200g/mL
Boiling Point 173°C to 174°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5300 to 1.532
Quantity 100 mL
Beilstein 18, 276
Specific Gravity 1.32
Merck Index 15, 4339
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether,toluene,methylene chloride,,dichloroethane,chlorobenzene,ethyl acetate,thf,and chloroform,decomposes in alcohol
Formula Weight 130.53
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Furoyl chloride, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
EINECSNumber 208-422-9
TSCA TSCA

RUO – Research Use Only