missing translation for 'onlineSavingsMsg'
Learn More

2-Furoic acid hydrazide, 98%

Catalog No. AC157160050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC157160050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC157160050 Supplier Thermo Scientific Chemicals Supplier No. 157160050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3326-71-4 | C5H6N2O2 | 126.115 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3326-71-4
MDL Number MFCD00003235
InChI Key SKTSVWWOAIAIKI-UHFFFAOYSA-N
Synonym 2-furoic acid hydrazide, 2-furoic hydrazide, 2-furohydrazide, furoylhydrazide, 2-furancarbohydrazide, 2-furoylhydrazide, furoic acid, hydrazide, 2-furoic acid, hydrazide, 2-furoylhydrazine, 2-furylcarbonylhydrazide
PubChem CID 18731
IUPAC Name furan-2-carbohydrazide
SMILES C1=COC(=C1)C(=O)NN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 76°C to 80°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H6N2O2
Quantity 5 g
Beilstein 18, 279
Molecular Weight (g/mol) 126.11
Formula Weight 126.11
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 2-Furoic acid hydrazide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
Harmful in contact with skin.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-046-2
RTECSNumber LV1925000
TSCA TSCA

RUO – Research Use Only