missing translation for 'onlineSavingsMsg'
Learn More

2-Furoic acid, 98%

Catalog No. AAA1145422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1145422 100 g
AAA1145436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1145422 Supplier Thermo Scientific Chemicals Supplier No. A1145422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 88-14-2 | C5H4O3 | 112.084 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 88-14-2
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.084
MDL Number MFCD00003238
InChI Key SMNDYUVBFMFKNZ-UHFFFAOYSA-N
Synonym 2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
PubChem CID 6919
ChEBI CHEBI:30845
IUPAC Name furan-2-carboxylic acid
SMILES C1=COC(=C1)C(=O)O

Specifications

Melting Point 127°C to 132°C
Boiling Point 230°C to 232°C (sublimation)
Flash Point 137°C (278°F)
Odor Odorless
Quantity 100 g
Beilstein 110149
Merck Index 14,4307
Formula Weight 112.08
Percent Purity 98%
Chemical Name or Material 2-Furoic acid
Hazard Category H315-H318-H335
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 201-803-0
RTECSNumber LV1763000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only