missing translation for 'onlineSavingsMsg'
Learn More

2-Furanmethanethiol, Analytical Standard, ≥98.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019369
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019369 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019369 Supplier Merck Emd Millipore Supplier No. 67319100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 98-02-2
Density 1.132 g/mL (at 25°C)
Physical Form Neat
Linear Formula C5H6OS
MDL Number MFCD00003254
Boiling Point 155°C
Synonym 2-Furfurylthiol; 2-Furylmethanethiol; Furfuryl mercaptan
Quantity 100 mg
Formula Weight 114.17
Percent Purity ≥98.0% (GC)
Grade Analytical Standard
Show More Show Less