missing translation for 'onlineSavingsMsg'
Learn More

2-Furancarboxyaldehyde, ACS, 98%, Spectrum™ Chemical

Catalog No. 18608216
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608216 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18608216 Supplier Spectrum Chemical Mfg Cor Supplier No. F1098500MLGL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 98-01-1
CAS Max % 1
Boiling Point 161.8°C (323.2°F)
Packaging Amber Glass Bottle
Quantity 500 mL
Residue after Evaporation 0.005
Molecular Weight (g/mol) 96.09
Percent Purity 0.98
Grade Reagent
Titratable Acid 0.02 meq/g
Physical Form Viscous Liquid or Low Melting Solid
Chemical Name or Material 2-Furancarboxyaldehyde
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.