missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorophénylacétone 97,0+%, TCI America™
Spécifications
Spécifications
| Color | Jaune |
| Boiling Point | 47°C |
| Quantity | 5 g |
| Formula Weight | 152.17 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquide |
| Chemical Name or Material | 2-Fluorophenylacetone |
Sécurité et manipulation
| TSCA | No |