missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenol, 98%

Catalog No. AAA1497922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1497922 100 g
AAA1497914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1497922 Supplier Thermo Scientific Chemicals Supplier No. A1497922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 367-12-4 | C6H5FO | 112.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 367-12-4
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.10
MDL Number MFCD00002155
InChI Key HFHFGHLXUCOHLN-UHFFFAOYSA-N
Synonym o-fluorophenol, phenol, 2-fluoro, phenol, o-fluoro, 1-fluoro-2-hydroxybenzene, 2-fluorohydroxybenzene, fluorophenol, unii-i9ow1nly9r, 2-fluoro-phenol, phenol, fluoro, ccris 1226
PubChem CID 9707
IUPAC Name 2-fluorophenol
SMILES OC1=CC=CC=C1F

Specifications

Melting Point 14°C to 16°C
Density 1.215
Boiling Point 150°C to 152°C
Flash Point 46°C (114°F)
Odor Phenol-like
Refractive Index 1.514
Quantity 100 g
UN Number UN2924
Beilstein 1905112
Formula Weight 112.1
Percent Purity 98%
Chemical Name or Material 2-Fluorophenol
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318-H226
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 206-681-2
RTECSNumber SL4530000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only