missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroisobutyric acid, 95%

Catalog No. AAH25837MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25837MD 250 mg
AAH2583703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25837MD Supplier Thermo Scientific Chemicals Supplier No. H25837MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 63812-15-7 | C4H7FO2 | 106.096 g/mol

2-Fluoroisobutyric acid chloride was synthesized from 2-fluoroisobutyric acid and oxalyl chloride. Reactant in the synthesis of 2-Fluoro-Acids, Esters, and amides From a-Dicyanoepoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoroisobutyric acid chloride was synthesized from 2-fluoroisobutyric acid and oxalyl chloride. Reactant in the synthesis of 2-Fluoro-Acids, Esters, and amides From a-Dicyanoepoxides.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Chemical Identifiers

CAS 63812-15-7
Molecular Formula C4H7FO2
Molecular Weight (g/mol) 106.096
MDL Number MFCD03452788
InChI Key NZDOWZQRNZLBOY-UHFFFAOYSA-N
Synonym 2-fluoroisobutyric acid, 2-fluoroisobutanoic acid, 2-fluoro-2-methyl-propanoic acid, 2-fluoro-2-methylpropionic acid, fluoroisobutyric acid, acmc-1b6kg, 2-methyl-2-fluoropropionic acid, 2-fluoro-2-methyl-propionic acid, 2-fluoranyl-2-methyl-propanoic acid, propanoic acid, 2-fluoro-2-methyl
PubChem CID 2782828
IUPAC Name 2-fluoro-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)F

Specifications

Boiling Point 57°C to 58°C (10 mmHg)
Refractive Index 1.385
Quantity 250 mg
UN Number UN2922
Solubility Information Soluble in water.
Formula Weight 106.1
Percent Purity 95%
Chemical Name or Material 2-Fluoroisobutyric acid
Hazard Category H301+H311-H314-H330-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only