missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorobiphenyl Solution, 2,000 μg/mL in dichloromethane, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental
Specifications
Specifications
| Packaging | Ampule |
| CAS | 321-60-8 |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 2,000 μg/mL in dichloromethane |