missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluorobenzyl Bromide 98.0+%, TCI America™
$53.90 - $146.31
Chemical Identifiers
| CAS | 446-48-0 |
|---|---|
| Molecular Formula | C7H6BrF |
| Molecular Weight (g/mol) | 189.027 |
| MDL Number | MFCD00000324 |
| InChI Key | FFWQLZFIMNTUCZ-UHFFFAOYSA-N |
| Synonym | 2-fluorobenzyl bromide, 1-bromomethyl-2-fluorobenzene, 2-fluorobenzylbromide, o-fluorobenzyl bromide, alpha-bromo-o-fluorotoluene, alpha-bromo-2-fluorotoluene, 2-fluoro-benzyl bromide, 1-bromomethyl-2-fluoro-benzene, benzene, 1-bromomethyl-2-fluoro, 2-fluoro-benzylbromide |
| PubChem CID | 67968 |
| IUPAC Name | 1-(bromomethyl)-2-fluorobenzene |
| SMILES | C1=CC=C(C(=C1)CBr)F |
Chemical Identifiers
| 446-48-0 | |
| 189.027 | |
| FFWQLZFIMNTUCZ-UHFFFAOYSA-N | |
| 67968 | |
| C1=CC=C(C(=C1)CBr)F |
| C7H6BrF | |
| MFCD00000324 | |
| 2-fluorobenzyl bromide, 1-bromomethyl-2-fluorobenzene, 2-fluorobenzylbromide, o-fluorobenzyl bromide, alpha-bromo-o-fluorotoluene, alpha-bromo-2-fluorotoluene, 2-fluoro-benzyl bromide, 1-bromomethyl-2-fluoro-benzene, benzene, 1-bromomethyl-2-fluoro, 2-fluoro-benzylbromide | |
| 1-(bromomethyl)-2-fluorobenzene |
Specifications
| 446-48-0 | |
| 115°C | |
| MFCD00000324 | |
| 3265 | |
| FFWQLZFIMNTUCZ-UHFFFAOYSA-N | |
| 1-(bromomethyl)-2-fluorobenzene | |
| 67968 | |
| ≥98.0% (GC) | |
| 2-Fluorobenzyl Bromide |
| Yellow | |
| C7H6BrF | |
| 5 g | |
| 2-fluorobenzyl bromide, 1-bromomethyl-2-fluorobenzene, 2-fluorobenzylbromide, o-fluorobenzyl bromide, alpha-bromo-o-fluorotoluene, alpha-bromo-2-fluorotoluene, 2-fluoro-benzyl bromide, 1-bromomethyl-2-fluoro-benzene, benzene, 1-bromomethyl-2-fluoro, 2-fluoro-benzylbromide | |
| C1=CC=C(C(=C1)CBr)F | |
| 189.027 | |
| 189.03 | |
| Liquid |
Safety and Handling
TSCA : No