accessibility menu, dialog, popup

UserName

FBr

CAS RN 446-48-0

IUPAC Name: 1-(bromomethyl)-2-fluorobenzene
Synonyms: 1-(bromomethyl)-2-fluorobenzene
Molecular Weight (g/mol): 189.03
Molecular Formula: C7H6BrF
InChi Key: FFWQLZFIMNTUCZ-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1CBr
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)