missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzyl Bromide 98.0+%, TCI America™

Supplier:  TCI America F020925G

 View more versions of this product

Catalog No. F020925G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Fluorobenzyl Bromide
Yellow
C7H6BrF
25 g
2-fluorobenzyl bromide, 1-bromomethyl-2-fluorobenzene, 2-fluorobenzylbromide, o-fluorobenzyl bromide, alpha-bromo-o-fluorotoluene, alpha-bromo-2-fluorotoluene, 2-fluoro-benzyl bromide, 1-bromomethyl-2-fluoro-benzene, benzene, 1-bromomethyl-2-fluoro, 2-fluoro-benzylbromide
C1=CC=C(C(=C1)CBr)F
189.027
189.03
Liquid
446-48-0
115°C
MFCD00000324
3265
FFWQLZFIMNTUCZ-UHFFFAOYSA-N
1-(bromomethyl)-2-fluorobenzene
67968
≥98.0% (GC)

Chemical Identifiers

446-48-0
189.027
FFWQLZFIMNTUCZ-UHFFFAOYSA-N
67968
C1=CC=C(C(=C1)CBr)F
C7H6BrF
MFCD00000324
2-fluorobenzyl bromide, 1-bromomethyl-2-fluorobenzene, 2-fluorobenzylbromide, o-fluorobenzyl bromide, alpha-bromo-o-fluorotoluene, alpha-bromo-2-fluorotoluene, 2-fluoro-benzyl bromide, 1-bromomethyl-2-fluoro-benzene, benzene, 1-bromomethyl-2-fluoro, 2-fluoro-benzylbromide
1-(bromomethyl)-2-fluorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions