missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzonitrile, 99%

Numéro de catalogue. AC250050250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC250050250 25 g Glass bottle
AC250050050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC250050250 Fournisseur Thermo Scientific Chemicals Code fournisseur 250050250
Il en reste null

CAS: 394-47-8 | C7H4FN | 121.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 394-47-8
Molecular Formula C7H4FN
Molecular Weight (g/mol) 121.11
MDL Number MFCD00001773
InChI Key GDHXJNRAJRCGMX-UHFFFAOYSA-N
Synonym o-fluorobenzonitrile, benzonitrile, 2-fluoro, o-cyanofluorobenzene, 2-fluorobenzonitril, 2-cyanofluorobenzene, benzonitrile, o-fluoro, fluorobenzonitrile, 2-fluorobenzenecarbonitrile, 2-fluoro-benzonitrile, 1-cyano-2-fluorobenzene
PubChem CID 67855
IUPAC Name 2-fluorobenzonitrile
SMILES FC1=CC=CC=C1C#N

Spécifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown to Colorless
Density 1.1100g/mL
Boiling Point 90°C (21.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.5063 to 1.5083
Quantity 25 g
Specific Gravity 1.11
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 121.12
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzonitrile
Afficher plus de résultats Afficher moins de résultats
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fre
EINECSNumber 206-897-7

RUO – Research Use Only