missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzenthiol 98,0+%, TCI America™

Catalog No. F04195G
Change view
Click to view available options
Quantité:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
F04195G 5 g
F041925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04195G Supplier TCI America Supplier No. F04195G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2557-78-0
Formule moléculaire C6H4FS
Poids moléculaire (g/mol) 127.16
Numéro MDL MFCD00041419
Clé InChI WJTZZPVVTSDNJJ-UHFFFAOYSA-M
Synonyme 2-fluorothiophenol, o-fluorothiophenol, o-fluorobenzenethiol, 2-fluorobenzene-1-thiol, fluorothiophenol, o-fluoro thiophenol, 2-fluoro-thiophenol, 2-fluoro-benzenethiol, 2-fluorobenzene thiol, pubchem5428
PubChem CID 520216
Nom de l’IUPAC (2-fluorophényl)sulfanure
SOURIRES FC1=CC=CC=C1[S-]

Specifications

Nom ou substance chimique 2-Fluorobenzenethiol
Couleur Jaune
Point d’ébullition 164 °C
Quantité 5 g
Numéro UN 3336
Poids de formule 128.16
Pourcentage de pureté ≥98,0% (GC)
Forme physique Liquide