missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroadenine 98.0+%, TCI America™

Catalog No. F0647200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0647200MG 200 mg
F06471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0647200MG Supplier TCI America Supplier No. F0647200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 700-49-2
Molecular Formula C5H4FN5
Molecular Weight (g/mol) 153.12
MDL Number MFCD01632749
InChI Key WKMPTBDYDNUJLF-UHFFFAOYSA-N
Synonym 2-fluoroadenine, 2-fluoro-6-aminopurine, 2-fluoro-1h-purin-6-amine, 2-fad, 2-fluoro-9h-purin-6-amine, 6-amino-2-fluoropurine, 2-fluoro-7 9 h-purin-6-ylamine, purine, 6-amino-2-fluoro, fluoroadenine, 1h-purin-6-amine, 2-fluoro
PubChem CID 12790
ChEBI CHEBI:72457
IUPAC Name 2-fluoro-7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC(F)=N1

Specifications

Quantity 200 mg
Formula Weight 153.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoroadenine
RTECSNumber AU6264700
TSCA No