missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroadenine, 97%

Catalog No. AC459160050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC459160050 5 g
AC459160010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC459160050 Supplier Thermo Scientific Chemicals Supplier No. 459160050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 700-49-2 | C5H4FN5 | 153.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 700-49-2
Molecular Formula C5H4FN5
Molecular Weight (g/mol) 153.12
MDL Number MFCD01632749
InChI Key WKMPTBDYDNUJLF-UHFFFAOYSA-N
Synonym 2-fluoroadenine, 2-fluoro-6-aminopurine, 2-fluoro-1h-purin-6-amine, 2-fad, 2-fluoro-9h-purin-6-amine, 6-amino-2-fluoropurine, 2-fluoro-7 9 h-purin-6-ylamine, purine, 6-amino-2-fluoro, fluoroadenine, 1h-purin-6-amine, 2-fluoro
PubChem CID 12790
ChEBI CHEBI:72457
IUPAC Name 2-fluoro-7H-purin-6-amine
SMILES NC1=C2NC=NC2=NC(F)=N1

Specifications

Melting Point >350.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 153.12
Percent Purity 97%
Chemical Name or Material 2-Fluoroadenine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

RUO – Research Use Only