missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. F08265G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08265G 5 g
F082625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08265G Supplier TCI America Supplier No. F08265G
Only null left

Chemical Identifiers

CAS 94239-04-0
Molecular Formula C6H3F4N
Molecular Weight (g/mol) 165.091
MDL Number MFCD03092901
InChI Key IZOIOCQPMHHDHN-UHFFFAOYSA-N
Synonym 2-fluoro-6-trifluoromethyl pyridine, 2-trifluoromethyl-6-fluoropyridine, pyridine, 2-fluoro-6-trifluoromethyl, 6-fluoro-2-trifluoromethyl pyridine, pubchem2403, acmc-209rqu, ksc488k3d, tpc-py087, 2-fluoro-6-trifluoromethyl py, 2-fluoro-6-rifluoromethylpyridine
PubChem CID 2736501
IUPAC Name 2-fluoro-6-(trifluoromethyl)pyridine
SMILES C1=CC(=NC(=C1)F)C(F)(F)F

Specifications

Color Colorless
Quantity 5 g
UN Number 1993
Formula Weight 165.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)pyridine