missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-nitrophénol 98,0+%, TCI America™

Catalog No. F06235G
Change view
Click to view available options
Quantité:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantité
F06235G 5 g
F06231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06235G Supplier TCI America Supplier No. F06235G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1526-17-6
Formule moléculaire C6H4FNO3
Poids moléculaire (g/mol) 157.10
Numéro MDL MFCD00042446
Clé InChI HIGRXCJEFUYRNW-UHFFFAOYSA-N
Synonyme 2-fluoro-6-nitrophenol, 6-fluoro-2-nitrophenol, phenol, 2-fluoro-6-nitro, 2-nitro-6-fluorophenol, 2-fluoro-6-nitro-phenol, 3-fluoro-2-hydroxynitrobenzene, fluoro-6-nitrophenol, 2, 2-fluoro-6-nitrophen, pubchem10654, 2-fluor-6-nitrophenol
PubChem CID 73710
Nom de l’IUPAC 2-fluoro-6-nitrophénol
SOURIRES OC1=C(F)C=CC=C1[N+]([O-])=O

Specifications

Nom ou substance chimique 2-Fluoro-6-nitrophenol
Point de fusion 92 °C
Couleur Jaune
Quantité 5 g
Poids de formule 157.10
Pourcentage de pureté ≥98,0% (GC)
Forme physique Poudre cristalline