missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitrobenzaldehyde 98.0+%, TCI America™

Catalog No. F06185G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06185G 5 g
F06181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F06185G Supplier TCI America Supplier No. F06185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27996-87-8
Molecular Formula C7H4FNO3
Molecular Weight (g/mol) 169.111
MDL Number MFCD00042298
InChI Key VVXFDFQEIRGULC-UHFFFAOYSA-N
Synonym fluoro nitrobenzaldehyde, 5-nitro-2-fluorobenzaldehyde, 4-fluoro-3-formylnitrobenzene, 2-fluoro-5-nitro-benzaldehyde, benzaldehyde, 2-fluoro-5-nitro, pubchem1459, acmc-209h0h, akos bb460, asischem d51510, 2-fluoro-5 nitrobenzaldehyde
PubChem CID 2734770
IUPAC Name 2-fluoro-5-nitrobenzaldehyde
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C=O)F

Specifications

Melting Point 59°C
Color Yellow
Quantity 5 g
Formula Weight 169.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-nitrobenzaldehyde