missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitroaniline, 98%

Catalog No. AC168391000
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168391000 100 g Glass bottle
AC168390050 5 g Glass bottle
AC168390250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC168391000 Supplier Thermo Scientific Chemicals Supplier No. 168391000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 369-36-8 | C6H5FN2O2 | 156.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 369-36-8
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.12
MDL Number MFCD00007652
InChI Key KJVBJICWGQIMOZ-UHFFFAOYSA-N
Synonym benzenamine, 2-fluoro-5-nitro, 2-fluoro-5-nitrophenylamine, 2-fluoro-5-nitro-phenylamine, 1-amino-2-fluoro-5-nitrobenzene, 2-fluoro-5-nitrobenzenamine, 2-fluoro-5-nitro aniline, 3-amino-4-fluoronitrobenzene, pubchem8504, acmc-1crtl, 5-nitro-2-fluoroaniline
PubChem CID 67785
IUPAC Name 2-fluoro-5-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])N)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 99°C to 103°C
Color Brown to Ochre-Yellow
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula FC6H3(NO2)NH2
Quantity 100 g
Beilstein 12, I, 355
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone,alcohol and toluene
Formula Weight 156.12
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-5-nitroaniline, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 206-720-3
TSCA TSCA

RUO – Research Use Only