missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitroaniline 98.0+%, TCI America™

Catalog No. F07985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F07985G 5 g
F07981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F07985G Supplier TCI America Supplier No. F07985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 369-35-7
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.116
MDL Number MFCD00034560
InChI Key LETNCFZQCNCACQ-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline
PubChem CID 101254
IUPAC Name 2-fluoro-4-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)N

Specifications

Melting Point 135°C
Color Yellow
Quantity 5 g
Formula Weight 156.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4-nitroaniline