missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-nitroaniline, 98%, Thermo Scientific™
Spécifications
Spécifications
| Nom ou substance chimique | 2-Fluoro-4-nitroaniline |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 97.5% min. (GC) |
| Formule linéaire | FC6H3(NO2)NH2 |
| Quantité | 5g |
| Poids de formule | 156.11 |
| Pourcentage de pureté | 98% |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position |
| EINECSNumber | 206-719-8 |
RUO – Research Use Only