missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-nitroaniline, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 457600250
| Quantity | 25g |
|---|
Chemical Identifiers
| 369-35-7 | |
| 156.11 | |
| 2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline | |
| 2-fluoro-4-nitroaniline |
| C6H5FN2O2 | |
| LETNCFZQCNCACQ-UHFFFAOYSA-N | |
| 101254 | |
| C1=CC(=C(C=C1[N+](=O)[O-])F)N |
Specifications
| 2-Fluoro-4-nitroaniline | |
| Authentic | |
| C6H5FN2O2 | |
| 25g | |
| LETNCFZQCNCACQ-UHFFFAOYSA-N | |
| 2-fluoro-4-nitroaniline | |
| 101254 | |
| 98% |
| 369-35-7 | |
| 97.5% min. (GC) | |
| FC6H3(NO2)NH2 | |
| 2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline | |
| C1=CC(=C(C=C1[N+](=O)[O-])F)N | |
| 156.11 | |
| 156.11 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
GHS Signal Word: Warning
EINECSNumber : 206-719-8
Promotions
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.