missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitroaniline, 98%, Thermo Scientific™

Catalog No. AC457600050
Change view
Click to view available options
Quantity:
5g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457600050 5g
AC457600250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC457600050 Supplier Thermo Scientific Chemicals Supplier No. 457600050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 369-35-7
Molecular Formula C6H5FN2O2
Molecular Weight (g/mol) 156.11
InChI Key LETNCFZQCNCACQ-UHFFFAOYSA-N
Synonym 2-fluoro-4-nitrobenzenamine, 2-fluoro-4-nitro-phenylamine, benzenamine, 2-fluoro-4-nitro, 2-fluoro-4-nitrophenylamine, 4-amino-3-fluoronitrobenzene, 2-fluoro-4-nitroaniline, pubchem8503, acmc-1bncp, 2-fluoro-4-niroaniline, 2-fluoro-4-nitro-aniline
PubChem CID 101254
IUPAC Name 2-fluoro-4-nitroaniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])F)N

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula FC6H3(NO2)NH2
Quantity 5g
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-nitroaniline
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position
EINECSNumber 206-719-8

RUO – Research Use Only