missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-hydroxybenzoic Acid 98.0+%, TCI America™

Catalog No. F05715G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05715G 5 g
F057125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05715G Supplier TCI America Supplier No. F05715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 65145-13-3
Molecular Formula C7H5FO3
Molecular Weight (g/mol) 156.11
MDL Number MFCD01310985
InChI Key NXWTWYULZRDBSA-UHFFFAOYSA-N
Synonym 2-fluoro-4-hydroxy-benzoic acid, benzoic acid, 2-fluoro-4-hydroxy, pubchem2608, acmc-1b9jp, 4-hydroxyfluorobenzoic acid, frinton fr-2445, ksc352q5p, 2-fluoro-4-hydroxybenzoicacid, 4-hydroxy-2-fluorobenzoic acid, 2-fluoro4-hydroxy-benzoic acid
PubChem CID 2783398
IUPAC Name 2-fluoro-4-hydroxybenzoic acid
SMILES OC(=O)C1=C(F)C=C(O)C=C1

Specifications

Melting Point 202°C
Color White-Yellow
Quantity 5 g
Formula Weight 156.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-4-hydroxybenzoic Acid