missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-3-nitropyridine 98.0+%, TCI America™

Catalog No. F09825G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09825G 5 g
F098225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F09825G Supplier TCI America Supplier No. F09825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1480-87-1 | C5H3FN2O2 | 142.09 g/mol

Chemical Identifiers

CAS 1480-87-1
Molecular Formula C5H3FN2O2
Molecular Weight (g/mol) 142.09
MDL Number MFCD03095068
InChI Key QDKIYDGHCFZBGC-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitro-pyridine, pyridine, 2-fluoro-3-nitro, pubchem1115, acmc-1bwlp, 2-fluor-3-nitropyridine, 2-fluoro-3-nitropyridine, ksc174g2n
PubChem CID 2762817
IUPAC Name 2-fluoro-3-nitropyridine
SMILES [O-][N+](=O)C1=C(F)N=CC=C1

Specifications

Melting Point 18°C
Boiling Point 110°C
Quantity 5 g
Formula Weight 142.09
Percent Purity ≥98.0% (GC)
Grade Laboratory
Physical Form Liquid
Chemical Name or Material 2-Fluoro-3-nitropyridine