missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Boiling Point | 140°C |
| Quantity | 200 mg |
| UN Number | 3272 |
| Formula Weight | 152.00 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Sécurité et manipulation
| TSCA | No |
| Recommended Storage | Refrigerator |