missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylthiophene, 99%
Description
It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
It is used as a heterocyclic building block. Also a maillard product used to provide a pleasant aroma to meat and coffee.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Density | 0.991 |
| Boiling Point | 133°C to 134°C |
| Flash Point | 21°C (69°F) |
| Odor | Stench |
| Refractive Index | 1.512 |
| Quantity | 100 g |
| UN Number | UN1993 |
| Beilstein | 105402 |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 112.19 |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319-H335 |
| Hazard Statement | GHS H StatementH225-H315-H319-H335Highly flammable liquid and vapour.Causes skin irritation.Causes serious eye irritation.May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 212-830-2 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only