missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylthiazole 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 148°C |
| Quantity | 5 g |
| UN Number | 1993 |
| Formula Weight | 113.18 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Ethylthiazole |
Safety and Handling
| TSCA | No |