missing translation for 'onlineSavingsMsg'
Learn More

2-Éthylimidazole 98,0+%, TCI America™

Numéro de catalogue. E023325G
Modifier l'affichage
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
E023325G 25 g
E0233500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. E023325G Fournisseur TCI America Code fournisseur E023325G

Identifiants chimiques

CAS 1072-62-4
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005192
InChI Key PQAMFDRRWURCFQ-UHFFFAOYSA-N
Synonym 2-ethylimidazole, 1h-imidazole, 2-ethyl, imidazole, 2-ethyl, 2-ethylimidazole 98+%, 2ez, 2-ethylimdazole, 2-ethyl imidazole, 2-ethyl-imidazole, pubchem15972
PubChem CID 66130
IUPAC Name 2-éthyl-1H-imidazole
SMILES CCC1=NC=CN1

Spécifications

Melting Point 83°C
Color Blanc-jaune
Quantity 25 g
Formula Weight 96.13
Percent Purity ≥98.0% (GC,T)
Physical Form Poudre cristalline
Chemical Name or Material 2-Ethylimidazole
EINECSNumber (5)-0382
TSCA Yes