missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Methacrylate (stabilized with MEHQ) 99.0+%, TCI America™

Catalog No. M059125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M059125ML 25 mL
M0591500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M059125ML Supplier TCI America Supplier No. M059125ML

Chemical Identifiers

CAS 688-84-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00009494
InChI Key WDQMWEYDKDCEHT-UHFFFAOYSA-N
Synonym 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
PubChem CID 12713
IUPAC Name 2-ethylhexyl 2-methylprop-2-enoate
SMILES CCCCC(CC)COC(=O)C(=C)C

Specifications

Color Colorless
Boiling Point 88°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1039
RTECSNumber OZ4630000
TSCA Yes