missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl methacrylate, 98%, stab. with 4-methoxyphenol

Catalog No. AA42303M4
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42303M4 20 L
AA42303AK 250 mL
AA42303K2 1 L
AA42303K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA42303M4 Supplier Thermo Scientific Chemicals Supplier No. 042303M4

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Monomer used in the preparation of polymethacrylate polymers | CAS: 688-84-6 | C12H22O2 | 198.306 g/mol

Monomer used in the preparation of polymethacrylate polymers

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 688-84-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00009494
InChI Key WDQMWEYDKDCEHT-UHFFFAOYSA-N
Synonym 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
PubChem CID 12713
IUPAC Name 2-ethylhexyl 2-methylprop-2-enoate
SMILES CCCCC(CC)COC(=O)C(=C)C

Specifications

Melting Point -50°C
Color Colorless
Density 0.885 g/mL
Boiling Point 218°C
Flash Point 92°C (198°F)
Odor Ether-like
Refractive Index 1.438
Quantity 20 L
Beilstein 1769420
Formula Weight 198.31
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl methacrylate, Stabilized with 4-methoxyphenol
Show More Show Less
Hazard Category H227-H315-H317-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c
EINECSNumber 211-708-6
RTECSNumber OZ4630000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only