missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl Diphenyl Phosphate 90.0+%, TCI America™

Catalog No. P1021500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1021500G 500 g
P102125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P1021500G Supplier TCI America Supplier No. P1021500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1241-94-7
Molecular Formula C20H27O4P
Molecular Weight (g/mol) 362.41
MDL Number MFCD00059949
InChI Key CGSLYBDCEGBZCG-UHFFFAOYNA-N
Synonym Octyl Diphenyl Phosphate, Phosphoric Acid Diphenyl 2-Ethylhexyl Ester, Phosphoric Acid 2-Ethylhexyl Diphenyl Ester, Phosphoric Acid Octyl Diphenyl Ester
PubChem CID 14716
IUPAC Name 2-ethylhexyl diphenyl phosphate
SMILES CCCCC(CC)COP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1

Specifications

Melting Point -30°C
Color Colorless
Boiling Point 232°C
Quantity 500 g
UN Number 3082
Formula Weight 362.41
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl Diphenyl Phosphate
EINECSNumber (3)-2520
RTECSNumber TC6125000
TSCA Yes