missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl 4-Hydroxybenzoate 98.0+%, TCI America™

Catalog No. H0506500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0506500G 500 g
H050625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0506500G Supplier TCI America Supplier No. H0506500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5153-25-3
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.338
MDL Number MFCD00016472
InChI Key VTIMKVIDORQQFA-UHFFFAOYSA-N
Synonym 2-Ethylhexylparaben, 4-Hydroxybenzoic Acid 2-Ethylhexyl Ester, 4-Hydroxybenzoic Acid Octyl Ester, Octyl 4-Hydroxybenzoate, Octylparaben
PubChem CID 107377
ChEBI CHEBI:34274
IUPAC Name 2-ethylhexyl 4-hydroxybenzoate
SMILES CCCCC(CC)COC(=O)C1=CC=C(C=C1)O

Specifications

Color Yellow
Boiling Point 270°C
Quantity 500 g
Formula Weight 250.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl 4-Hydroxybenzoate
EINECSNumber (3)-1585
TSCA Yes